Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POMP
Entrez Gene ID
510314
Gene Name
proteasome maturation protein
Gene Aliases
-
Chromosome Location
Chr.12: 31471714 - 31482858 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034376.2 NP_001029548.1 4 - 5 301 76
GenBank mRNA BC102689.1 - 4 - 5 302 76

View Full Product Details