Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO6
Entrez Gene ID
513023
Gene Name
F-box protein 6
Gene Aliases
-
Chromosome Location
Chr.16: 42003245 - 42021104 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034433.1 NP_001029605.1 3 - 4 375 109
GenBank mRNA BC104546.1 - 3 - 4 375 109
BT030507.1 - 3 - 4 387 109

View Full Product Details