Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC6A
Entrez Gene ID
514979
Gene Name
C-type lectin domain containing 6A
Gene Aliases
-
Chromosome Location
Chr.5: 101642073 - 101659403 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034479.1 NP_001029651.1 3 - 4 387 100
GenBank mRNA DQ176046.1 - 4 - 5 489 100
DQ176047.1 - 3 - 4 387 100

View Full Product Details