Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A34
Entrez Gene ID
515553
Gene Name
solute carrier family 25 member 34
Gene Aliases
-
Chromosome Location
Chr.16: 51819920 - 51824310 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034497.2 NP_001029669.1 1 - 2 553 72
GenBank mRNA BC102816.1 - 1 - 2 554 72

View Full Product Details