Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYB5R1
Entrez Gene ID
516287
Gene Name
cytochrome b5 reductase 1
Gene Aliases
-
Chromosome Location
Chr.16: 79860414 - 79865628 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034518.1 NP_001029690.1 8 - 9 789 73
GenBank mRNA BC104584.1 - 8 - 9 789 73

View Full Product Details