Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLUL1
Entrez Gene ID
516801
Gene Name
clusterin like 1
Gene Aliases
-
Chromosome Location
Chr.24: 35518976 - 35545065 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034520.1 NP_001029692.1 7 - 8 1292 119
GenBank mRNA AY643094.1 - 7 - 8 1292 119

View Full Product Details