Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAR1A
Entrez Gene ID
517171
Gene Name
secretion associated Ras related GTPase 1A
Gene Aliases
-
Chromosome Location
Chr.28: 26382360 - 26395931 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034521.2 NP_001029693.1 4 - 5 358 115
GenBank mRNA BC102443.1 - 4 - 5 359 115

View Full Product Details