Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1QBP
Entrez Gene ID
518321
Gene Name
complement C1q binding protein
Gene Aliases
-
Chromosome Location
Chr.19: 26114946 - 26120376 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034527.2 NP_001029699.1 3 - 4 587 100
GenBank mRNA BC102464.1 - 3 - 4 588 100

View Full Product Details