The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034582.2 | NP_001029754.1 | 2 - 3 | 372 | 76 |
GenBank mRNA | BC102487.1 | - | 2 - 3 | 373 | 76 |