Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCCHC17
Entrez Gene ID
538816
Gene Name
zinc finger CCHC-type containing 17
Gene Aliases
-
Chromosome Location
Chr.2: 122295336 - 122347024 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034652.2 NP_001029824.1 4 - 5 301 80
GenBank mRNA BC102395.1 - 4 - 5 303 80
BT025465.1 - 3 - 4 196 80

View Full Product Details