Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB2
Entrez Gene ID
539244
Gene Name
ankyrin repeat and SOCS box containing 2
Gene Aliases
-
Chromosome Location
Chr.21: 58530359 - 58579232 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034669.1 NP_001029841.1 2 - 3 391 93
GenBank mRNA BC104497.1 - 2 - 3 391 93

View Full Product Details