Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSPAN33
Entrez Gene ID
539284
Gene Name
tetraspanin 33
Gene Aliases
-
Chromosome Location
Chr.4: 93068920 - 93088748 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034672.2 NP_001029844.1 6 - 7 844 72
GenBank mRNA BC103364.1 - 6 - 7 845 72

View Full Product Details