Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAN
Entrez Gene ID
540457
Gene Name
RAN, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.17: 46283208 - 46288136 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034705.2 NP_001029877.1 4 - 5 305 103
GenBank mRNA BC102559.1 - 4 - 5 306 103
BC151426.1 - 4 - 5 311 103

View Full Product Details