Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11H2orf42
Entrez Gene ID
540909
Gene Name
chromosome 11 C2orf42 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 68473608 - 68513219 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034714.2 NP_001029886.1 6 - 7 1293 67
GenBank mRNA BC103343.1 - 6 - 7 1294 67

View Full Product Details