Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THY1
Entrez Gene ID
614712
Gene Name
Thy-1 cell surface antigen
Gene Aliases
-
Chromosome Location
Chr.15: 29981019 - 29986913 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034765.1 NP_001029937.1 3 - 4 438 82
GenBank mRNA BC104530.1 - 3 - 4 438 82

View Full Product Details