Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM18
Entrez Gene ID
616554
Gene Name
transmembrane protein 18
Gene Aliases
-
Chromosome Location
Chr.8: 112473017 - 112477327 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034793.1 NP_001029965.1 1 - 2 104 94
GenBank mRNA BC103183.1 - 1 - 2 104 94

View Full Product Details