Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM18
Entrez Gene ID
616554
Gene Name
transmembrane protein 18
Gene Aliases
-
Chromosome Location
Chr.8: 112473017 - 112477327 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034793.1 NP_001029965.1 4 - 5 370 67
GenBank mRNA BC103183.1 - 4 - 5 370 67

View Full Product Details