Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC2
Entrez Gene ID
616748
Gene Name
trafficking protein particle complex 2
Gene Aliases
-
Chromosome Location
Chr.X: 129796070 - 129808212 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034796.1 NP_001029968.1 3 - 4 311 97
GenBank mRNA BC102418.1 - 3 - 4 311 97

View Full Product Details