Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSRB1
Entrez Gene ID
618441
Gene Name
methionine sulfoxide reductase B1
Gene Aliases
SEPX1
Chromosome Location
Chr.25: 1502594 - 1507246 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034810.2 NP_001029982.2 3 - 4 475 112
GenBank mRNA BC105188.1 - 3 - 4 423 112
CK836798.1 - 3 - 4 475 112

View Full Product Details