Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIMM50
Entrez Gene ID
505489
Gene Name
translocase of inner mitochondrial membrane 50
Gene Aliases
-
Chromosome Location
Chr.18: 49160944 - 49168043 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035024.1 NP_001030196.1 5 - 6 377 63
GenBank mRNA BC102990.1 - 5 - 6 377 63
BT026310.1 - 5 - 6 389 63

View Full Product Details