Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FSCN1
Entrez Gene ID
507342
Gene Name
fascin actin-bundling protein 1
Gene Aliases
-
Chromosome Location
Chr.25: 38748351 - 38757827 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035045.1 NP_001030217.1 2 - 3 1116 119
GenBank mRNA BC105199.1 - 2 - 3 1116 119

View Full Product Details