Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SHQ1
Entrez Gene ID
507486
Gene Name
SHQ1, H/ACA ribonucleoprotein assembly factor
Gene Aliases
-
Chromosome Location
Chr.22: 28835678 - 28939926 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035049.2 NP_001030221.1 9 - 10 1141 152
GenBank mRNA BC105240.1 - 9 - 10 1142 152

View Full Product Details