Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PALMD
Entrez Gene ID
509823
Gene Name
palmdelphin
Gene Aliases
-
Chromosome Location
Chr.3: 43536425 - 43596104 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035079.2 NP_001030251.1 7 - 8 1755 59
GenBank mRNA BC105234.1 - 7 - 8 1756 59

View Full Product Details