Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTAQ1
Entrez Gene ID
510463
Gene Name
N-terminal glutamine amidase 1
Gene Aliases
C14H8orf32, C8orf32, WDYHV1
Chromosome Location
Chr.14: 16424582 - 16443170 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035091.1 NP_001030263.1 2 - 3 259 101
GenBank mRNA BC102447.1 - 2 - 3 259 101

View Full Product Details