Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRSF6
Entrez Gene ID
507828
Gene Name
serine and arginine rich splicing factor 6
Gene Aliases
SFRS6
Chromosome Location
Chr.13: 72096697 - 72102392 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035272.1 NP_001030349.1 1 - 2 252 69
GenBank mRNA BC107553.1 - 1 - 2 252 69

View Full Product Details