Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDF2L1
Entrez Gene ID
517962
Gene Name
stromal cell derived factor 2 like 1
Gene Aliases
-
Chromosome Location
Chr.17: 72112697 - 72114638 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035323.1 NP_001030400.1 2 - 3 455 69
GenBank mRNA BC102528.1 - 2 - 3 455 69

View Full Product Details