Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOSC3
Entrez Gene ID
533245
Gene Name
exosome component 3
Gene Aliases
-
Chromosome Location
Chr.8: 61883370 - 61888055 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035397.2 NP_001030474.1 3 - 4 642 101
GenBank mRNA BC102436.1 - 3 - 4 643 101

View Full Product Details