Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTFR1
Entrez Gene ID
535449
Gene Name
mitochondrial fission regulator 1
Gene Aliases
-
Chromosome Location
Chr.14: 30038845 - 30099662 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035412.2 NP_001030489.1 2 - 3 176 153
GenBank mRNA BC102798.1 - 2 - 3 178 153

View Full Product Details