Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASP6
Entrez Gene ID
538409
Gene Name
caspase 6
Gene Aliases
-
Chromosome Location
Chr.6: 15694142 - 15714982 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035419.2 NP_001030496.1 2 - 3 83 92
GenBank mRNA BC102309.1 - 2 - 3 84 92

View Full Product Details