Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPST2
Entrez Gene ID
540183
Gene Name
tyrosylprotein sulfotransferase 2
Gene Aliases
-
Chromosome Location
Chr.17: 66190150 - 66244471 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035429.2 NP_001030506.1 6 - 7 1314 68
GenBank mRNA BC103334.1 - 6 - 7 1315 68

View Full Product Details