Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IPMK
Entrez Gene ID
615135
Gene Name
inositol polyphosphate multikinase
Gene Aliases
-
Chromosome Location
Chr.26: 765339 - 819755 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035462.1 NP_001030539.1 3 - 4 686 109
GenBank mRNA BC102793.1 - 3 - 4 686 109

View Full Product Details