Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIP6
Entrez Gene ID
615869
Gene Name
thyroid hormone receptor interactor 6
Gene Aliases
-
Chromosome Location
Chr.25: 35785398 - 35789235 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035469.1 NP_001030546.1 7 - 8 1302 63
GenBank mRNA BC104544.1 - 7 - 8 1302 63

View Full Product Details