Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL4
Entrez Gene ID
507154
Gene Name
mitochondrial ribosomal protein L4
Gene Aliases
-
Chromosome Location
Chr.7: 14771719 - 14781062 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037450.2 NP_001032527.1 2 - 3 171 124
GenBank mRNA BC108102.1 - 2 - 3 172 124

View Full Product Details