Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UXT
Entrez Gene ID
525680
Gene Name
ubiquitously expressed prefoldin like chaperone
Gene Aliases
-
Chromosome Location
Chr.X: 86023688 - 86030097 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037471.2 NP_001032548.1 6 - 7 449 70
GenBank mRNA BC108205.1 - 6 - 7 450 70

View Full Product Details