Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDCA7
Entrez Gene ID
614893
Gene Name
cell division cycle associated 7
Gene Aliases
-
Chromosome Location
Chr.2: 23340521 - 23353702 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037488.2 NP_001032565.1 1 - 2 129 129
GenBank mRNA BC108118.1 - 1 - 2 130 129

View Full Product Details