Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EEF1G
Entrez Gene ID
326581
Gene Name
eukaryotic translation elongation factor 1 gamma
Gene Aliases
-
Chromosome Location
Chr.29: 40917049 - 40926643 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040487.2 NP_001035577.1 2 - 3 193 112
GenBank mRNA AB098935.1 - 1 - 1 170 112
AB099020.1 - 1 - 1 187 112
AB099085.1 - 1 - 2 170 112
BC102691.1 - 2 - 3 194 112
BT025451.1 - 1 - 2 170 112

View Full Product Details