Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EHHADH
Entrez Gene ID
518852
Gene Name
enoyl-CoA hydratase and 3-hydroxyacyl CoA dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.1: 81877820 - 81932666 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075780.2 NP_001069248.2 2 - 3 214 93
GenBank mRNA BC102238.1 - 2 - 3 201 93

View Full Product Details