Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19H17orf64
Entrez Gene ID
530096
Gene Name
chromosome 19 C17orf64 homolog
Gene Aliases
C17orf64
Chromosome Location
Chr.19: 12521668 - 12532887 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075929.2 NP_001069397.2 3 - 4 329 73
GenBank mRNA BC109987.1 - 2 - 3 206 73

View Full Product Details