The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075979.2 | NP_001069447.1 | 3 - 4 | 440 | 118 |
GenBank mRNA | BC108214.1 | - | 3 - 4 | 442 | 118 |