The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076427.1 | NP_001069895.1 | 1 - 2 | 199 | 122 |
GenBank mRNA | BC102811.1 | - | 1 - 2 | 199 | 122 |