Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2C87
Entrez Gene ID
616593
Gene Name
cytochrome P450, family 2, subfamily C, polypeptide 87
Gene Aliases
-
Chromosome Location
Chr.26: 16144354 - 16208922 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076427.1 NP_001069895.1 6 - 7 989 66
GenBank mRNA BC102811.1 - 6 - 7 989 66

View Full Product Details