Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
QDPR
Entrez Gene ID
618084
Gene Name
quinoid dihydropteridine reductase
Gene Aliases
-
Chromosome Location
Chr.6: 112919767 - 112939911 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076492.2 NP_001069960.1 5 - 6 585 127
GenBank mRNA BC102193.1 - 5 - 6 586 127

View Full Product Details