Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL21
Entrez Gene ID
618609
Gene Name
mitochondrial ribosomal protein L21
Gene Aliases
-
Chromosome Location
Chr.29: 46258417 - 46268365 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076514.2 NP_001069982.1 6 - 7 577 105
GenBank mRNA BC109734.1 - 6 - 7 578 105

View Full Product Details