Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TERF1
Entrez Gene ID
519474
Gene Name
telomeric repeat binding factor 1
Gene Aliases
-
Chromosome Location
Chr.14: 36382250 - 36425376 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076865.2 NP_001070333.2 1 - 2 344 72
GenBank mRNA BC104542.1 - 1 - 2 344 72
DV828419.1 - 1 - 2 328 72
EV614740.1 - 1 - 2 338 72

View Full Product Details