Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG12
Entrez Gene ID
767903
Gene Name
autophagy related 12
Gene Aliases
-
Chromosome Location
Chr.10: 4680210 - 4702517 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076982.1 NP_001070450.1 2 - 3 308 99
GenBank mRNA BC102225.1 - 2 - 3 308 99

View Full Product Details