Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2H2orf88
Entrez Gene ID
767914
Gene Name
chromosome 2 C2orf88 homolog
Gene Aliases
smAKAP
Chromosome Location
Chr.2: 6097453 - 6178350 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076991.2 NP_001070459.1 1 - 2 38 98
GenBank mRNA BC102650.1 - 1 - 2 40 98

View Full Product Details