Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDHAF4
Entrez Gene ID
768071
Gene Name
succinate dehydrogenase complex assembly factor 4
Gene Aliases
C9H6orf57
Chromosome Location
Chr.9: 9683227 - 9729000 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077128.2 NP_001070596.1 2 - 3 245 80
GenBank mRNA BC111629.1 - 2 - 3 247 80

View Full Product Details