Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HBA
Entrez Gene ID
512439
Gene Name
hemoglobin, alpha 2
Gene Aliases
HBA2, alpha globin
Chromosome Location
Chr.25: 216496 - 217264 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077422.3 NP_001070890.2 1 - 2 134 96
GenBank mRNA BC102940.1 - 1 - 2 135 96
BC133477.1 - 1 - 2 134 96

View Full Product Details