Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DECR2
Entrez Gene ID
768256
Gene Name
2,4-dienoyl-CoA reductase 2
Gene Aliases
-
Chromosome Location
Chr.25: 388100 - 396057 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078122.1 NP_001071590.1 8 - 9 995 88
GenBank mRNA BC103065.1 - 8 - 9 995 88

View Full Product Details