Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YWHAQ
Entrez Gene ID
768311
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta
Gene Aliases
-
Chromosome Location
Chr.11: 87862015 - 87893173 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078127.2 NP_001071595.1 4 - 5 711 73
GenBank mRNA BC102840.1 - 4 - 5 712 73
BC151414.1 - 3 - 4 633 73
BT030749.2 - 3 - 4 665 73

View Full Product Details